1-(Dimethylisopropylsilyl)imidazole 95%
Supplier: Apollo Scientific
OR938220-1G
OR938220-250MG
APOSOR938220-1GEA
386
EUR
APOSOR938220-1G
APOSOR938220-250MG
1-(Dimethylisopropylsilyl)imidazole 95%
1-(Dimethylisopropylsilyl)imidazole
Formula:
C₈H₁₆N₂Si MW: 168.32 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059891 CAS Number: 81452-04-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...