4-Methoxy-2,5-dimethylbenzaldehyde 95%
Supplier: Apollo Scientific
OR938285-10G OR938285-1G OR938285-5G OR938285-250MG OR938285-500MG
APOSOR938285-10GEA 437 EUR
APOSOR938285-10G APOSOR938285-1G APOSOR938285-5G APOSOR938285-250MG APOSOR938285-500MG
4-Methoxy-2,5-dimethylbenzaldehyde 95%
4-Methoxy-2,5-dimethylbenzaldehyde
Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003343
CAS Number: 6745-75-1