cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%
Supplier: Apollo Scientific
OR938498-10G OR938498-1G OR938498-5G
APOSOR938498-10GEA 1010 EUR
APOSOR938498-10G APOSOR938498-1G APOSOR938498-5G
cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid 95%
cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid
Formula: C₈H₁₄O₃
MW: 158.20 g/mol
Storage Temperature: Ambient
CAS Number: 73094-35-6