4-Bromo-2,6-xylidine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-2,6-dimethylaniline
OR9386-100G OR9386-1KG OR9386-500G
APOSOR9386-100GEA 27.7 EUR
APOSOR9386-100G APOSOR9386-1KG APOSOR9386-500G
4-Bromo-2,6-xylidine 98%
4-Bromo-2,6-xylidine
Formula: C₈H₁₀BrN
Boiling Pt: 120 °C/0.2mmHg
Melting Pt: 48…51 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007826
CAS Number: 24596-19-8
EINECS: 246-337-9