trans-2,3-Dichloro-1,4-dioxane 95%
Supplier: Apollo Scientific
Synonyms: trans-2,3-Dichloro-1,4-diethylene Dioxide
OR938993-1G OR938993-25G OR938993-5G
APOSOR938993-1GEA 125 EUR
APOSOR938993-1G APOSOR938993-25G APOSOR938993-5G
trans-2,3-Dichloro-1,4-dioxane 95%
trans-2,3-Dichloro-1,4-dioxane
Formula: C₄H₆Cl₂O₂
MW: 156.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191421
CAS Number: 3883-43-0