cis-1,3-Cyclohexanediol 95%
Supplier: Apollo Scientific
Synonyms: cis-1,3-Dihydroxycyclohexane
OR939088-1G OR939088-250MG
APOSOR939088-1GEA 632 EUR
APOSOR939088-1G APOSOR939088-250MG
cis-1,3-Cyclohexanediol 95%
cis-1,3-Cyclohexanediol
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Storage Temperature: Ambient
CAS Number: 823-18-7