6-Bromocaproic acid 98+%
Supplier: Apollo Scientific
Danger
Synonyms: 6-Bromohexanoic acid
OR9393-100G OR9393-25G OR9393-5G
APOSOR9393-100GEA 26.1 EUR
APOSOR9393-100G APOSOR9393-25G APOSOR9393-5G
6-Bromocaproic acid 98+%
6-Bromocaproic acid
Formula: BrH₂C(CH₂)₄COOH
MW: 195.06 g/mol
Boiling Pt: 165…170 °C/20mmHg
Melting Pt: 32…35 °C
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00004422
CAS Number: 4224-70-8
EINECS: 224-176-5
UN: 3261
ADR: 8,II