1-Methyl-1,2-epoxycyclohexane 95%
Supplier: Apollo Scientific
Synonyms: 1-Methyl-7-oxabicyclo[4.1.0]heptane, 1,2-Epoxy-1-methylcyclohexane
OR939738-5G
APOSOR939738-5GEA 186 EUR
APOSOR939738-5G
1-Methyl-1,2-epoxycyclohexane 95%
1-Methyl-1,2-epoxycyclohexane
Formula: C₇H₁₂O
MW: 112.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846121
CAS Number: 1713-33-3