O-Phenylhydroxylamine hydrochloride 95%
Supplier: Apollo Scientific
Synonyms: Hydroxylamine, O-phenyl-, hydrochloride, Phenoxyamine hydrochloride, N-phenoxyamine hydrochloride
OR940013-1G OR940013-5G
APOSOR940013-1GEA 56.7 EUR
APOSOR940013-1G APOSOR940013-5G
O-Phenylhydroxylamine hydrochloride 95%
O-Phenylhydroxylamine hydrochloride
Formula: C₆H₈ClNO
MW: 145.586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043271
CAS Number: 6092-80-4