5-Bromo-2-methoxy-N-methylbenzenesulphonamide 95%
Supplier: Apollo Scientific
Warning
OR9401-1G OR9401-5G OR9401-25G
APOSOR9401-1GEA 114 EUR
APOSOR9401-1G APOSOR9401-5G APOSOR9401-25G
5-Bromo-2-methoxy-N-methylbenzenesulphonamide 95%
5-Bromo-2-methoxy-N-methylbenzenesulphonamide
Formula: C₈H₁₀BrNO₃S
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD06808832
CAS Number: 871269-17-9