5-Bromo-N-tert-butyl-2-methoxybenzenesulphonamide 98%
Supplier: Apollo Scientific
OR9403-1G OR9403-5G
APOSOR9403-1GEA 139 EUR
APOSOR9403-1G APOSOR9403-5G
5-Bromo-N-tert-butyl-2-methoxybenzenesulphonamide 98%
5-Bromo-N-tert-butyl-2-methoxybenzenesulphonamide
Formula: C₁₁H₁₆BrNO₃S
Melting Pt: 208…212 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08056376
CAS Number: 871269-15-7