3-(2-Ethylhexyloxy)-1,2-propanediol
Supplier: Apollo Scientific
Synonyms: Glycerol alpha-(2-Ethylhexyl) Ether, 3-((2-Ethylhexyl)oxy)propane-1,2-diol
OR942257-1KG OR942257-500G OR942257-250G OR942257-25G OR942257-100G
APOSOR942257-1KGEA 714 EUR
APOSOR942257-1KG APOSOR942257-500G APOSOR942257-250G APOSOR942257-25G APOSOR942257-100G
3-(2-Ethylhexyloxy)-1,2-propanediol
3-(2-Ethylhexyloxy)-1,2-propanediol
Formula: C₁₁H₂₄O₃
MW: 204.31 g/mol
Density: 0.96 g/cm³ (25 °C)
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD03790878
CAS Number: 70445-33-9