1,1-Cyclohexanediacetic anhydride 97%
Supplier: Apollo Scientific
OR942282-100G OR942282-25G
APOSOR942282-100GEA 142 EUR
APOSOR942282-100G APOSOR942282-25G
1,1-Cyclohexanediacetic anhydride 97%
1,1-Cyclohexanediacetic anhydride
Formula: C₁₀H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00040292
CAS Number: 1010-26-0