1,1-Cyclopentanediacetic anhydride 95%
Supplier: Apollo Scientific
Synonyms: 3,3-Tetramethyleneglutaric Anhydride, 8-Oxaspiro[4.5]decane-7,9-dione, 8-Oxaspiro[4.5]-7,9-decanedione
OR942532-1G OR942532-5G
APOSOR942532-1GEA 50.9 EUR
APOSOR942532-1G APOSOR942532-5G
1,1-Cyclopentanediacetic anhydride 95%
1,1-Cyclopentanediacetic anhydride
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006678
CAS Number: 5662-95-3