(R)-tert-Butyl (1-aminopropan-2-yl)carbamate 97%
Supplier: Apollo Scientific
Synonyms: (R)-2-N-Boc-propane-1,2-diamine, tert-Butyl (R)-1-aminopropan-2-ylcarbamate
OR943505-1G OR943505-500MG
APOSOR943505-1GEA 654 EUR
APOSOR943505-1G APOSOR943505-500MG
(R)-tert-Butyl (1-aminopropan-2-yl)carbamate 97%
(R)-tert-Butyl (1-aminopropan-2-yl)carbamate
Formula: C₈H₁₈N₂O₂
MW: 174.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11112229
CAS Number: 100927-10-4