2-Chloropyridine
Supplier: Apollo Scientific
Danger
OR9447-100G OR9447-500G
APOSOR9447-100GEA 111 EUR
APOSOR9447-100G APOSOR9447-500G
2-Chloropyridine
2-Chloropyridine
Formula: C₅H₄ClN
MW: 113.55 g/mol
Boiling Pt: 166 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00006228
CAS Number: 109-09-1
EINECS: 203-646-3
UN: 2822
ADR: 6.1,II