2-Pyridinethiol
Supplier: Apollo Scientific
Warning
Synonyms: Pyridine-2-thiol, 2-Mercaptopyridine
OR9448-100G OR9448-25G OR9448-5G
APOSOR9448-100GEA 78.3 EUR
APOSOR9448-100G APOSOR9448-25G APOSOR9448-5G
2-Pyridinethiol
2-Pyridinethiol
Formula: C₅H₅NS
MW: 111.17 g/mol
Melting Pt: 127…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006285
CAS Number: 2637-34-5
EINECS: 220-131-9