4-tert-Butyl-1-bromobenzene, Technical Grade
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-4-tert-butylbenzene, 2-(4-Bromophenyl)-2-methylpropane, 1-Bromo-4-(tert-butyl)benzene, 4-Bromo-tert-butylbenzene
OR9457-100G OR9457-500G OR9457-50G OR9457-1KG
APOSOR9457-100GEA 36.4 EUR
APOSOR9457-100G APOSOR9457-500G APOSOR9457-50G APOSOR9457-1KG
4-tert-Butyl-1-bromobenzene, Technical Grade
4-tert-Butyl-1-bromobenzene
Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 228…232 °C
Melting Pt: 13…16 °C
Density: 1.229 g/cm³ (25 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00000108
CAS Number: 3972-65-4
EINECS: 223-599-2