1,5-Dimethyl-1H-pyrazole-4-carbonitrile 95%
Supplier: Apollo Scientific
OR945825-1G OR945825-250MG
APOSOR945825-1GEA 183 EUR
APOSOR945825-1G APOSOR945825-250MG
1,5-Dimethyl-1H-pyrazole-4-carbonitrile 95%
1,5-Dimethyl-1H-pyrazole-4-carbonitrile
Formula: C₆H₇N₃
MW: 121.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04969708
CAS Number: 856860-16-7