4-(2-Methoxyethoxy)benzaldehyde 98%
Supplier: Apollo Scientific
OR946440-10G OR946440-1G OR946440-5G
APOSOR946440-10GEA 502 EUR
APOSOR946440-10G APOSOR946440-1G APOSOR946440-5G
4-(2-Methoxyethoxy)benzaldehyde 98%
4-(2-Methoxyethoxy)benzaldehyde
Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08144121
CAS Number: 36824-00-7