2-Bromo-5-methoxy-4-methylaniline 98%
Supplier: Apollo Scientific
Warning
OR947349-10G OR947349-1G OR947349-5G
APOSOR947349-10GEA 514 EUR
APOSOR947349-10G APOSOR947349-1G APOSOR947349-5G
2-Bromo-5-methoxy-4-methylaniline 98%
2-Bromo-5-methoxy-4-methylaniline
Formula: C₈H₁₀BrNO
MW: 216.0751 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11841053
CAS Number: 532440-88-3