2-Nitronaphthalene 98%
Supplier: Apollo Scientific
OR948091-1G OR948091-25G OR948091-5G
APOSOR948091-1GEA 453 EUR
APOSOR948091-1G APOSOR948091-25G APOSOR948091-5G
2-Nitronaphthalene 98%
2-Nitronaphthalene
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 314 °C
Melting Pt: 65…79 °C
Storage Temperature: Refrigerator
CAS Number: 581-89-5
UN: 2538
ADR: 4.1,III