3-Chloro-5-methoxyphenylboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: 3-Chloro-5-methoxybenzeneboronic acid
OR9487-1G OR9487-25G OR9487-5G OR9487-250MG
APOSOR9487-1GEA 43.7 EUR
APOSOR9487-1G APOSOR9487-25G APOSOR9487-5G APOSOR9487-250MG
3-Chloro-5-methoxyphenylboronic acid
3-Chloro-5-methoxyphenylboronic acid
Formula: C₇H₈BClO₃
MW: 186.40 g/mol
Melting Pt: 185…189 °C
Storage Temperature: Ambient
MDL Number: MFCD03095046
CAS Number: 915201-07-9