5'-O-(4,4'-Dimethoxytrityl)thymidine 98%
Supplier: Apollo Scientific
Warning
Synonyms: DMT-T, 1-((2R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, 5'-O-Dimethoxytrityl-deoxythymidine
OR9495-100G OR9495-25G OR9495-5G
APOSOR9495-100GEA 423 EUR
APOSOR9495-100G APOSOR9495-25G APOSOR9495-5G
5'-O-(4,4'-Dimethoxytrityl)thymidine 98%
5'-O-(4,4'-Dimethoxytrityl)thymidine
Formula: C₃₁H₃₂N₂O₇
MW: 544.60 g/mol
Melting Pt: 151 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010113
CAS Number: 40615-39-2