(2E,4E,6E)-2,7-Dimethylocta-2,4,6-trienedial 95%
Supplier: Apollo Scientific
OR949640-1G OR949640-5G
APOSOR949640-1GEA 219 EUR
APOSOR949640-1G APOSOR949640-5G
(2E,4E,6E)-2,7-Dimethylocta-2,4,6-trienedial 95%
(2E,4E,6E)-2,7-Dimethylocta-2,4,6-trienedial
Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11113099
CAS Number: 5056-17-7