3-Methylcyclohexanol 95%
Supplier: Apollo Scientific
Synonyms: 3-Methylhexalin
OR949642-25G OR949642-500G OR949642-5G
APOSOR949642-25GEA 0 EUR
APOSOR949642-25G APOSOR949642-500G APOSOR949642-5G
3-Methylcyclohexanol 95%
3-Methylcyclohexanol
Formula: C₇H₁₄O
MW: 114.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001446
CAS Number: 591-23-1