3-Nitro-2,6-lutidine
Supplier: Apollo Scientific
Danger
Synonyms: 2,6-Dimethyl-3-nitropyridine, 3-Nitro-2,6-lutidine
OR9504-25G OR9504-5G OR9504-100G
APOSOR9504-25GEA 74.1 EUR
APOSOR9504-25G APOSOR9504-5G APOSOR9504-100G
3-Nitro-2,6-lutidine
3-Nitro-2,6-lutidine
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 26…31 °C
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00023503
CAS Number: 15513-52-7
EINECS: 239-543-5
UN: 1325
ADR: 4.1,III