2-Isopropenylthiazole 95%

Supplier: Apollo Scientific

OR950629-1G OR950629-5G
APOSOR950629-1GEA 270 EUR
APOSOR950629-1G APOSOR950629-5G
2-Isopropenylthiazole 95%
2-Isopropenylthiazole
Formula: C₆H₇NS
MW: 125.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00671547
CAS Number: 13816-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR