3-(3-Methylphenoxy)propionic acid 98%
Supplier: Apollo Scientific
OR951180-1G OR951180-5G
APOSOR951180-1GEA 161 EUR
APOSOR951180-1G APOSOR951180-5G
3-(3-Methylphenoxy)propionic acid 98%
3-(3-Methylphenoxy)propionic acid
Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00642411
CAS Number: 25173-36-8