2-Chloro-N-(2-hydroxypyridin-3-yl)acetamide 95%
Supplier: Apollo Scientific
OR951370-1G OR951370-250MG
APOSOR951370-1GEA 406 EUR
APOSOR951370-1G APOSOR951370-250MG
2-Chloro-N-(2-hydroxypyridin-3-yl)acetamide 95%
2-Chloro-N-(2-hydroxypyridin-3-yl)acetamide
Formula: C₇H₇ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12922757
CAS Number: 112777-30-7