2-Amino[1,8]naphthyridine-3-carbonitrile 98%
Supplier: Apollo Scientific
OR951392-1G OR951392-5G
APOSOR951392-1GEA 226 EUR
APOSOR951392-1G APOSOR951392-5G
2-Amino[1,8]naphthyridine-3-carbonitrile 98%
2-Amino[1,8]naphthyridine-3-carbonitrile
Formula: C₉H₆N₄
MW: 170.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659014
CAS Number: 15935-95-2