3-Amino-2,6-dimethylpyridine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Dimethylpyridin-3-amine
OR9514-100G OR9514-25G OR9514-5G
APOSOR9514-100GEA 163 EUR
APOSOR9514-100G APOSOR9514-25G APOSOR9514-5G
3-Amino-2,6-dimethylpyridine 98%
3-Amino-2,6-dimethylpyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 230 °C
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD01646409
CAS Number: 3430-33-9
EINECS: 222-332-7
UN: 2811
ADR: 6.1,III