2-(4-Methoxyphenyl)-2-methylpropanenitrile 95%
Supplier: Apollo Scientific
OR951453-1G OR951453-5G OR951453-250MG OR951453-500MG
APOSOR951453-1GEA 91.5 EUR
APOSOR951453-1G APOSOR951453-5G APOSOR951453-250MG APOSOR951453-500MG
2-(4-Methoxyphenyl)-2-methylpropanenitrile 95%
2-(4-Methoxyphenyl)-2-methylpropanenitrile
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00229868
CAS Number: 5351-07-5