7-Chloropyrazolo[1,5-a]pyrimidine 98%
Supplier: Apollo Scientific
OR951521-1G OR951521-5G
APOSOR951521-1GEA 101 EUR
APOSOR951521-1G APOSOR951521-5G
7-Chloropyrazolo[1,5-a]pyrimidine 98%
7-Chloropyrazolo[1,5-a]pyrimidine
Formula: C₆H₄ClN₃
MW: 153.5700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04035684
CAS Number: 58347-49-2