5-(Chloromethyl)-3-methyl-1,2,4-oxadiazole 97%
Supplier: Apollo Scientific
OR951851-1G OR951851-5G
APOSOR951851-1GEA 94.4 EUR
APOSOR951851-1G APOSOR951851-5G
5-(Chloromethyl)-3-methyl-1,2,4-oxadiazole 97%
5-(Chloromethyl)-3-methyl-1,2,4-oxadiazole
Formula: C₄H₅ClN₂O
MW: 132.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08445357
CAS Number: 1192-81-0