7-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%
Supplier: Apollo Scientific
Warning
OR952561-1G OR952561-25G OR952561-5G
APOSOR952561-1GEA 21.9 EUR
APOSOR952561-1G APOSOR952561-25G APOSOR952561-5G
7-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%
7-Bromo-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formula: C₉H₁₁BrClN
MW: 248.5500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094742
CAS Number: 220247-73-4