3-Aminopiperidin-2-one 96%
Supplier: Apollo Scientific
Synonyms: 3-Amino-2-piperidinone, 3-Amino-2-piperidone, 2-Amino-delta-valerolactam
OR953224-1G OR953224-25G OR953224-5G OR953224-0.8G
APOSOR953224-1GEA 60.9 EUR
APOSOR953224-1G APOSOR953224-25G APOSOR953224-5G APOSOR953224-0.8G
3-Aminopiperidin-2-one 96%
3-Aminopiperidin-2-one
Formula: C5H10N2O
MW: 114.1457 g/mol
Boiling Pt: 125…135 °C
Melting Pt: 49…52 °C
Storage Temperature: Freezer
MDL Number: MFCD06809610
CAS Number: 1892-22-4
Order Now
Specification Test Results
Purity 96%