Ethyl-4-formyl-1H-pyrrole-2-carboxylate 95%
Supplier: Apollo Scientific
Synonyms: Ethyl 4-formylpyrrole-2-carboxylate, Ethyl 4-formyl-1H-pyrrole-2-carboxylate
OR953465-1G OR953465-5G
APOSOR953465-1GEA 43.5 EUR
APOSOR953465-1G APOSOR953465-5G
Ethyl-4-formyl-1H-pyrrole-2-carboxylate 95%
Ethyl-4-formyl-1H-pyrrole-2-carboxylate
Formula: C₈H₉NO₃
MW: 167.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04122505
CAS Number: 7126-57-0