4-Bromo-o-toluic acid
Supplier: Apollo Scientific
Danger
Synonyms: 4-Bromo-2-methylbenzoic acid
OR9537-100G OR9537-25G OR9537-500G
APOSOR9537-100GEA 78.3 EUR
APOSOR9537-100G APOSOR9537-25G APOSOR9537-500G
4-Bromo-o-toluic acid
4-Bromo-o-toluic acid
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00040905
CAS Number: 68837-59-2
EINECS: 272-437-7
UN: 2811
ADR: 6.1,III