3-Bromo-5-chloro-1,2,4-thiadiazole
Supplier: Apollo Scientific
Warning
OR9538-10G OR9538-1G OR9538-5G
APOSOR9538-10GEA 277 EUR
APOSOR9538-10G APOSOR9538-1G APOSOR9538-5G
3-Bromo-5-chloro-1,2,4-thiadiazole
3-Bromo-5-chloro-1,2,4-thiadiazole
Formula: C₂BrClN₂S
MW: 199.45 g/mol
Boiling Pt: 192…194 °C
Melting Pt: 24…25 °C
Storage Temperature: Ambient
MDL Number: MFCD00799287
CAS Number: 37159-60-7