Isoxazolo[5,4-b]pyridin-3-ylamine 98%
Supplier: Apollo Scientific
Synonyms: Isoxazolo[5,4-b]pyridin-3-amine
OR953834-1G OR953834-5G
APOSOR953834-1GEA 122 EUR
APOSOR953834-1G APOSOR953834-5G
Isoxazolo[5,4-b]pyridin-3-ylamine 98%
Isoxazolo[5,4-b]pyridin-3-ylamine
Formula: C₆H₅N₃O
MW: 135.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04117798
CAS Number: 92914-74-4