4-Chlorobenzofuro[3,2-d]pyrimidine
Supplier: Apollo Scientific
Warning
OR953913-1G OR953913-250MG OR953913-5G
APOSOR953913-1GEA 58 EUR
APOSOR953913-1G APOSOR953913-250MG APOSOR953913-5G
4-Chlorobenzofuro[3,2-d]pyrimidine
4-Chlorobenzofuro[3,2-d]pyrimidine
Formula: C₁₀H₅ClN₂O
MW: 204.6100 g/mol
Melting Pt: 146 °C
Storage Temperature: Ambient
MDL Number: MFCD00763943
CAS Number: 39876-88-5