1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one 98%
Supplier: Apollo Scientific
Synonyms: 1-EBIO, 1-Ethyl-1H-benzo[d]imidazol-2(3H)-one, 1-Ethylbenzimidazolinone
OR954133-1G OR954133-5G
APOSOR954133-1GEA 43.5 EUR
APOSOR954133-1G APOSOR954133-5G
1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one 98%
1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one
Formula: C₉H₁₀N₂O
MW: 162.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005715
CAS Number: 10045-45-1