4-Methoxypyridine-2-carbonitrile 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-methoxypicolinonitrile
OR9544-1G OR9544-5G
APOSOR9544-1GEA 79.9 EUR
APOSOR9544-1G APOSOR9544-5G
4-Methoxypyridine-2-carbonitrile 95%
4-Methoxypyridine-2-carbonitrile
Formula: C₇H₆N₂O
MW: 134.1400 g/mol
Melting Pt: 118…121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08062857
CAS Number: 36057-44-0
UN: 3439
ADR: 6.1,III