3-(4-Methoxyphenyl)Pyrazole
Supplier: Apollo Scientific
Warning
Synonyms: 3-(4-Methoxyphenyl)-1H-pyrazole
OR9552-250MG OR9552-1G
APOSOR9552-250MGEA 21.9 EUR
APOSOR9552-250MG APOSOR9552-1G
3-(4-Methoxyphenyl)Pyrazole
3-(4-Methoxyphenyl)Pyrazole
Formula: C₁₀H₁₀N₂O
MW: 174.203 g/mol
Boiling Pt: 158 °C/0.6mmHg
Melting Pt: 130…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00112008
CAS Number: 27069-17-6