Methyl-2-bromoisonicotinate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-bromoisonicotinate
OR9560-1G OR9560-5G
APOSOR9560-1GEA 21.9 EUR
APOSOR9560-1G APOSOR9560-5G
Methyl-2-bromoisonicotinate 98%
Methyl 2-bromoisonicotinate
Formula: C₇H₆BrNO₂
MW: 216.0320 g/mol
Melting Pt: 35…37 °C
Storage Temperature: Ambient
MDL Number: MFCD03791265
CAS Number: 26156-48-9