Methyl-6-bromonicotinate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 6-bromonicotinate
OR9561-25G OR9561-5G
APOSOR9561-25GEA 66.7 EUR
APOSOR9561-25G APOSOR9561-5G
Methyl-6-bromonicotinate 97%
Methyl 6-bromonicotinate
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 106…110 °C
Storage Temperature: Ambient
MDL Number: MFCD04972371
CAS Number: 26218-78-0