Nitrocyclopentane 95%

Supplier: Apollo Scientific

OR956241-1G OR956241-5G
APOSOR956241-1GEA 139 EUR
APOSOR956241-1G APOSOR956241-5G
Nitrocyclopentane 95%
Nitrocyclopentane
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 40
Storage Temperature: Ambient
MDL Number: MFCD00001362
CAS Number: 2562-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR