2-Bromoisonicotinamide 97%

Supplier: Apollo Scientific

Synonyms: 2-Bromopyridine-4-carboxamide

OR9565-1G OR9565-25G OR9565-5G
APOSOR9565-1GEA 85.7 EUR
APOSOR9565-1G APOSOR9565-25G APOSOR9565-5G
2-Bromoisonicotinamide 97%
2-Bromoisonicotinamide
Formula: C₆H₅BrN₂O
MW: 201.02 g/mol
Melting Pt: 188…191 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234156
CAS Number: 29840-73-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR