4-Chloro-3-methyl-1H-pyrazole
Supplier: Apollo Scientific
Warning
OR956908-1G OR956908-5G
APOSOR956908-1GEA 79.9 EUR
APOSOR956908-1G APOSOR956908-5G
4-Chloro-3-methyl-1H-pyrazole
4-Chloro-3-methyl-1H-pyrazole
Formula: C₄H₅ClN₂
MW: 116.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01693485
CAS Number: 15878-08-7